Fluke 700HTP-2 Hydraulic Test Pump, 0 to 10,000 psi
Representative image only.
Fluke Mfr # FLUKE-700HTP-2 – Item # EW-16100-38
Available for sale only in the United States and Canada
  • Generate pressure up to 10,000 psi with accurate and reliable handhelds
  • 1 year warranty
$1,626.99USD / Each
sign in for your price.
$204.00 - $277.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Min Pressure0 psi
  • ConnectionsTwo 1/4\" NPT(F) ports
  • DescriptionHydraulic Test Pump, 0 to 10,000 psi
  • Warranty1 Year
1 Year Warranty
Key Features
  • Generate pressure up to 10,000 psi with accurate and reliable handhelds
  • 1 year warranty
More About this Item
The Fluke 700HTP-2 is a hand operated hydraulic pressure pump designed to generate calibration pressures up to 10,000 psi/690 bar. Designed for use with Fluke-700 series high pressure modules such as the Fluke-700P09, 700P29, 700P30 and 700P31. Ideal for calibration when connected to a Fluke high pressure module via the reference port. The 700HTP-2 uses distilled water or mineral-based hydraulic oil as a source for calibration of high pressure input devices such as pressure transmitters or pressure gauges. Can also be used with Fluke 717 pressure calibrators.