Fluke 437-II 3-Phase 400 Hz Power Quality Kit

Representative image only.
Fluke Mfr # FLUKE-437-II – Item # EW-20043-13
Quickly identify power problems, manage, and reduce energy waste
  • Energy loss calculator for pinpoint true system energy losses in dollars
  • Power inverter efficiency allows simultaneous measurement of AC output power and DC input power for power electronics systems using optional DC clamp
  • Troubleshoot real-time - analyzing trends using the cursors and zoom tools
  • Highest safety rating in the industry - 600 V CAT IV / 1000 V CAT III rated for use at the service entrance
  • Automatic trending ensures that every measurement is always automatically recorded, without any set-up
  • System monitors up to ten power quality parameters on one screen according to EN50160 power quality standard
  • Fully class-A compliant - analyzers conduct tests according to the stringent international IEC 61000-4-30 Class-A standard
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Min AC Voltage (Voltage)1
  • Max AC Voltage (Voltage)1000
  • Min AC Current (amps)0.5
  • Max AC Current (amps)6000
  • Min DC Voltage (Voltage)1
  • Max DC Voltage (Voltage)1000
  • Min DC Current (amps)0.5
  • Max DC Current (amps)6000
  • Power Factor Range0 to 1
  • HarmonicsHarmonics volts, THD, harmonic amps, K factor amps, harmonic watts, THD watts, K factor watts, interharmonic amps, Vrms, Arms (relative to fundamental or to total RMS)
  • Watts (AC/DC) Range6000 MW (max)
  • Memory1 hour up to 1 year, user selectable (default setting 7 days)
  • Sample Rate200 kS/s on each channel simultaneously
  • Display127 x 88 mm (6\" diagonal) LCD; 320 x 240 pixels
  • Battery TypeLi-ion battery; 28 Wh (up to 7 hours continuous use)
  • Height (in)13
  • Width (in)12 1/4
  • Depth (in)2 1/2
  • Height (cm)33
  • Width (cm)31.1
  • Depth (cm)63.5
  • Description3-Phase 400 Hz Power Quality Kit
  • Warranty2 Years
Two year warranty

Key Features

  • Energy loss calculator for pinpoint true system energy losses in dollars
  • Power inverter efficiency allows simultaneous measurement of AC output power and DC input power for power electronics systems using optional DC clamp
  • Troubleshoot real-time - analyzing trends using the cursors and zoom tools
  • Highest safety rating in the industry - 600 V CAT IV / 1000 V CAT III rated for use at the service entrance
  • Automatic trending ensures that every measurement is always automatically recorded, without any set-up
  • System monitors up to ten power quality parameters on one screen according to EN50160 power quality standard
  • Fully class-A compliant - analyzers conduct tests according to the stringent international IEC 61000-4-30 Class-A standard
  • 2 year warranty

More About this Item

Available for Sale in the United States and Canada only

The new Fluke 430 Series II three-phase power quality analyzers help you locate, predict, prevent, and troubleshoot problems in three-phase and single-phase power distribution systems. Units feature an exclusive energy loss algorithm, Unified Power Measurement, that measures and quantifies energy losses due to harmonics and unbalance issues, allowing you to pinpoint the origin of energy waste within a system. Troubleshooting is faster with on-screen display of trends and captured events, even while background recording continues. The IEC standards for flicker, harmonics and power quality, are all built-in taking the guesswork out of power quality.

All models feature volt, amp, hertz, dips and swells, harmonics, power and energy, energy loss calculator, unbalance, monitor, power inverter efficiency, and inrush capabilities.

Additional features include event waveform capture, flicker, transients, mains signaling, and power wave. Model 437-II only provides 400 Hz measurement capability.



What's included: Test lead and alligator clip set (TL430), four 24" (61 cm L) flexible current probes (i430), software, 8 GB SD card, USB cable, hard carrying case with rollers (437-II), power adapter (BC430), international plug adapter set, and Li-ion battery (BP290).

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat