Fluke 233 True-RMS Digital Multimeter with Remote Display

Representative image only.
Fluke Mfr # FLUKE-233 – Item # EW-20034-40
Available for sale in the United States and Canada only Remote display allows you to be in two places at once
  • Removable magnetic display can be conveniently mounted where it is easily seen
  • Unique design makes this meter perfect for hazardous areas, cleanrooms, and more
  • Measures
  • True RMS ac voltage and current
  • Measure up to 1000 V ac and dc

$332.99USD / Each
$369.99USD / Each

$94.00USD / Each

for an additional
$94.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • DC Min (Volts)0.0001
  • DC Max (Volts)1000
  • DC Voltage Accuracy0.25% 2
  • DC Min (amps)0.0001
  • DC Max (amps)10
  • DC Current Accuracy1.0% 3
  • AC Min (Volts)0.0001
  • AC Max (Volts)1000
  • Ac Voltage Accuracy1.0% 3
  • AC Min (amps)0.0001
  • AC Max (amps)10
  • AC Current Accuracy1.5% 3
  • Resistance Min (Ω)0.1
  • Resistance Max (MΩ)40
  • Capacitance Min (nF)1000
  • Capacitance Max (mF)9999
  • Frequency Min (Hz)0.1
  • Frequency Max (KHz)50
  • Safety RatingCAT III 1000V, CAT IV 600V
  • Power3 x AA battery main body, 2 AA batteries display
  • DescriptionTrue-RMS Digital Multimeter with Remote Display

Includes

holster, test leads, alligator clips, 80BK-A temperature probe, CD-ROM, three AA batteries (main body), and two AA batteries (display).

Key Features

  • Removable magnetic display can be conveniently mounted where it is easily seen
  • Unique design makes this meter perfect for hazardous areas, cleanrooms, and more
  • Measures
  • True RMS ac voltage and current
  • Measure up to 1000 V ac and dc
  • Measure up to 10 A (20 A for 30 seconds)
  • 10,000 µF capacitance range
  • Frequency to 50 kHz
  • Built in thermometer conveniently allows you to take temperatures readings without having to carry a separate instrument
  • Resistance, continuity and diode test
  • Min/Max and average recording to capture variations automatically
  • Features
  • Wireless technology allows the display to be carried up to 30 ft. away from the point of measurement for added flexibility
  • The removable magnetic display can be conveniently mounted where it is easily seen
  • Make measurements without holding the meter to improve visual focus on probes and augment safe electrical measurements
  • Low-power 802.15.4 wireless technology does not interfere with measurement accuracy
  • Use as a conventional multimeter when the display is connected
  • Auto power off maximizes battery life
  • Radio transmitter automatically turns off when the display is connected to the meter
  • True-rms ac voltage and current for accurate measurements on non-linear signals
  • Record signal fluctuations using the Min/Max function
  • Test continuity and diodes
  • Easy to read display with large digits and bright backlight
  • Battery life approximately 200 hrs

More About this Item

This True RMS multimeter with remote display eliminates the dilemma of trying to take measurements in hard-to-reach places, or making measurements in machines or panels that are physically separated from a limit or isolator switch. The low power wireless technology allows the display to be carried up to 33 ft (10 m) away from the point of measurement for added flexibility, without interfering with the measurement.

Unit allows you to make measurements without holding the meter to improve visual focus on probes and augment safe electrical measurements. The radio transmitter automatically turns off when the display is connected to the meter allowing you to use as a conventional multimeter and maximizing battery life. Additional features include built-in thermometer, min/max and average, resistance, continuity and diode test, and an easy-to-read bright backlit display for dim or poorly lit conditions.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat