Flir Industrial Acoustic Imaging Camera
Representative image only.
Flir Mfr # Si124 – Item # EW-23029-11
Identify efficiency loss and potential failures up to 10 times faster than traditional methods
  • Locate costly leaks in noisy environments
  • Instantly view the leak rate and estimated yearly energy loss
  • Extend compressor life by eliminating wasted output
  • Identify partial discharge type to improve reliability of electrical systems
  • Recognize corona discharge, even in the daytime
  • Lightweight camera can be operated with one hand
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Decibel (Db) Low Range (dB)0
  • Decibel (Db) High Range (dB)120
  • OutputWi-Fi 2.4 GHz and 5 GHz IEEE 802.11.b/g/n/ac wireless LAN
  • Data LoggingYes
  • Memory32 GB SD card, non-removable and 8 GB USB mass storage, provided with device
  • DisplayResistive touchscreen
  • DescriptionIndustrial Acoustic Imaging Camera
  • Warranty2 year
Two year warranty
Includes
Camera, battery, battery cable, battery charger, battery pouch, camera hand strap, camera pouch, printed documentation, and USB memory stick.
Key Features
  • Locate costly leaks in noisy environments
  • Instantly view the leak rate and estimated yearly energy loss
  • Extend compressor life by eliminating wasted output
  • Identify partial discharge type to improve reliability of electrical systems
  • Recognize corona discharge, even in the daytime
  • Lightweight camera can be operated with one hand
  • 2 year warranty
More About this Item
This acoustic imager is designed to visually show pressurized leaks in compressed air systems and display partial discharge problems in high-voltage electrical systems. This technology can help utility, manufacturing, and engineering professionals. Built with 124 microphones and a frequency range that covers audible and ultrasound (2 kHz to 31 kHz), the Si124 sees through background noise commonly found in industrial environments to produce precise acoustic imagery. The acoustic image is overlaid in real time on top of a digital camera picture, which lets the user pinpoint the source of the sound and classify problems. Adopting the FLIR Si124 as part of a regular maintenance routine, professionals can help utilities keep the power flowing and manufacturing operations going.