Battery life>4 hours at 25°C (68°F) with typical use
Battery2 Rechargeable Lithium-ion (included)
DescriptionGas Thermal Imaging Camera with MSX® and 25° High-Range (HR), 9.5-12 µm Lens
Includes
Infrared camera with lens, power supply for battery charger, power supply 15 W/3 A, printed documentation, SD card (8 GB), USB 2.0 A to USB Type-C cable, USB Type-C to HDMI and PD adapter, USB Type-C to USB Type-C cable (USB 2.0 standard), lens cap strap, lens cleaning cloth, neck strap, small eyecup, battery (2x), battery charger, hard transport case, lens cap front, lens cap front and rear (only for extra lenses).
Key Features
Gas-specific, spectral filtered lenses can be easily changed out in the field
Temperature and gas measurement features can be used simultaneously
One tap on the screen automatically adjusts contrast between the gas and the background environment
High Sensitivity Mode (HSM) amplifies plume movement for easier gas detection
Data reporting is simplified through built-in voice annotation, GPS tagging, customizable work folders, and Wi-Fi connection
Includes 25° high-range lens
2 year warranty
More About this Item
This uncooled infrared camera measures temperature and visualizes gas emissions to accurately diagnose electrical and mechanical faults. The ergonomic design has a 180° rotating optical block, touch-screen LCD, and an eyepiece for use in direct sunlight. The camera can be used with multiple lenses that are interchangeable.
The lenses are spectrally filtered from 7 to 8.5 µm and 9.5 to 12 µm, respectively. The low-range (LR) lens are used for detecting methane, nitrous oxide, propane, sulfur dioxide, and refrigerants R-134a or R-152a. The high-range (HR) lens are for detecting sulfur hexafluoride, ammonia, and ethylene. The lenses allow the camera to scan components quickly, focusing on areas with small leaks that may be missed with a traditional TVA (Toxic Vapor Analyzer) or "sniffer". The efficiency of this leak detection protects the environment, lessens product losses, and provides a safer working environment.