Fisher Research Labs M-101 Rebar Locator W-Probe Case And Harness
Representative image only.
Fisher Research Labs Mfr # M-101 – Item # EW-20001-54
  • 2 year warranty
$1,149.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRebar Locator W-Probe Case And Harness
  • Warranty2 Years
Two year warranty
Key Features
  • 2 year warranty
More About this Item
This metal detector was designed specially for finding reinforcing bars and other metal objects embedded in concrete, conduits, PK nails, wiring, wall studs and other metallic objects. Users may expect accuracy to 1/4'' of the center line of the rebar.
Turn the instrument on and only the penetration control need be adjusted for most operations, allowing maximum penetration depth for metals.
A preset circuit eliminates ground effects caused by highly mineralized materials and false signals due to loop movement.
The detector can also effectively locate small metal items carried on or within a person and is substantially more sensitive than the average low cost metal detectors used at airports, so that a given metal item can be found at a greater distance from the search loop itself.
Searches in water may be up to the control box. The search loop and cord are waterproof.
FEATURES
  • Detects all metals
  • Accurately pinpoints steel reinforcing bars, or ''rebars'' in concrete;
  • (typically 8-9'' depth for 1/2'' diameter rebar)
  • Detects intersections of rebar or mesh
  • Audio and visual indications of nearby metal
  • Rugged construction
  • Operates on universally available 9V batteries
  • Quick tune ready for operation in seconds
  • Fully waterproof search loop
  • VLF, (Very Low Frequency), operation