Extech 601500 Standard pH Electrode, Glass; 12 x 160 mm, BNC
Representative image only.
Extech Mfr # 601500 – Item # EW-53026-42
  • BNC connector works with most meter brands
  • Sealed gelled reference filling solution and detachable bulb guard protect electrode
  • Full range 0 to 14.0 pH measurements and 0 to 80°C temperature compatibility
  • BNC pH connector
  • 6 month warranty
$42.56USD / Each
$47.29USD / Each
Include Buffers Today!
Oakton® pH Buffer Solution Packs
$39.00 - $405.00USD / Pkg of 3
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeAccessory; Probes and Sensors
  • DescriptionStandard pH Electrode, Glass; 12 x 160 mm
  • Warranty6 month
6 Month Warranty
Key Features
  • BNC connector works with most meter brands
  • Sealed gelled reference filling solution and detachable bulb guard protect electrode
  • Full range 0 to 14.0 pH measurements and 0 to 80°C temperature compatibility
  • BNC pH connector
  • 6 month warranty
More About this Item
Replace pH electrodes when they can no longer hold calibration, dry out and cannot be rehydrated, or if the housing cracks.