This item is now discontinued. We recommend the following as suggested replacements: EW-30009-26
Extech 480823 Field Meter Electromagnetic
Representative image only.
Extech Mfr # 480823 – Item # EW-20018-41
  • Large 1/2'' LCD (1999 count) display of Electromagnetic Field (EMF) level in milliGauss or microTesla
  • Accurate measurements to 4% over a measuring range of 0.1 to 199.9mGauss (0.01 to 19.99µTesla)
  • ELF frequency bandwidth of 30 to 300Hz
  • Single axis - sampling 2.5 times per second
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionField Meter Electromagnetic
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • Large 1/2'' LCD (1999 count) display of Electromagnetic Field (EMF) level in milliGauss or microTesla
  • Accurate measurements to 4% over a measuring range of 0.1 to 199.9mGauss (0.01 to 19.99µTesla)
  • ELF frequency bandwidth of 30 to 300Hz
  • Single axis - sampling 2.5 times per second