Explosion-proof NEMA Type C-Face Motor, 3/4 Hp, 208-230/460 VAC
Representative image only.
Item # EW-71007-35
For Class 1 Group C and D, Class 2 Group F and G locations
  • Automatic overload protection stops and restarts motor in case of overload or overheating
  • UL-listed file no. E-12044
PRODUCT OPTIONS
$1,005.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Motor RPM (rpm)1800
  • Motor Frame Size56C
  • Motor Enclosure TypeTEFC
  • Power (VAC)208 to 230/ 460
  • Power (amps)1.9-3.0/1.5
  • Power (Phase)3
  • Power Conduit Box ConnectionYes
  • Length (in)13.94
  • Length (cm)35.41
  • Motor hp3/4
  • DescriptionExplosion-proof Three-phase NEMA Type C-face Motor, 3/4 Hp, 208-230/460 VAC
Key Features
  • Automatic overload protection stops and restarts motor in case of overload or overheating
  • UL-listed file no. E-12044
More About this Item
Motors are TEFC (models 70007-00 to -12 are TENV) and include an explosion-proof conduit box (shipped loose).