Eurotherm COMMS BOARDMBUS EIA422
Representative image only.
Eurotherm Mfr # SUB24/EIA422 – Item # EW-94470-33
Highly stable with advanced programmable features.
  • 1 year warranty
$206.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The 2400 series are high stability controllers with an extensive range of options. Either PID, on/off or motorized valve control can be configured, satisfying both electrical and gas heating applications. Dual PID settings, advanced ‘one shot’ and continuously adaptive tuning are available to optimize control performance. These controllers employ universal inputs with Instant Accuracy™ and accept a range of plug-in modules for heating, cooling, analog retransmission, second process value input and remote setpoint. High speed Modbus® RTU, Profibus® DP, DeviceNet™, and Ei-Bisynch ASCII digital communications provide easy connection to PLC™, PC based supervisory control and data logging systems. Multi-zone programming can be implemented using PDSIO® retransmission to deliver a master setpoint to one, two, or three client controllers, with holdback from any client if the temperature deviates from the setpoint by more than a set value.