Eppendorf 940002109 Electroporation Buffer for Eukaryotic Cells, Iso-osmolar, 100 mL
Representative image only.
Eppendorf Mfr # 940002109 – Item # EW-36205-62
Provides quality transfection results
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)100
  • ColorClear
  • DescriptionElectroporation Buffer for Eukaryotic Cells, Iso-osmolar, 100 mL
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The best possible transfection results are obtained by using the original buffers from Eppendorf®, which have been tested for sterility as well as for the absence of mycoplasma, endotoxins, and pyrogens, and have undergone a thorough cytotoxicity test.
The iso-osmolar buffer 36205-62 is for electrofusion of animal cells. The buffer optimizes your cell fusion parameters by only requiring a few cells in your chamber.