Eppendorf 3124000121 Single-Channel Pipette, Blue; Fixed 1000 uL
Representative image only.
Eppendorf Mfr # 3124000121 – Item # EW-24505-31
Provide high reproducibility with your dispensing and reduce the fatigue associated with constant repetitive thumb and arm movements.
  • Requires up to 50% less pipetting force, designed to minimize stress and fatigue
  • The ultra light pipette is designed to fit comfortably in your hand
  • Made with industry leading materials for exceptional durability
  • 3 year warranty
PRODUCT OPTIONS
$380.00USD / Each
$89.50 - $162.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Sugggested alternative
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)1000
  • Max Volume (mL)1
  • Max Dispensing (mL)1
  • Color CodeBlue
  • DescriptionSingle-Channel Pipette, Blue; Fixed 1000 uL
  • Warranty3 year
3 Year Warranty
Key Features
  • Requires up to 50% less pipetting force, designed to minimize stress and fatigue
  • The ultra light pipette is designed to fit comfortably in your hand
  • Made with industry leading materials for exceptional durability
  • 3 year warranty
More About this Item
The Research Plus was optimized for form and function. The ultra lightweight pipette meets the highest needs for precision and accuracy- combined with ultimate ergonomics and increased flexibility. The spring loaded tip cone achieves optimal tighntess and minimal attachment forces-doing most of the work for you. Make your volume adjustments with just one hand- only a few turns will get you to your desired volume. The four-digit volume display is easy to view. Dissassemble your pipettor with ease- the quick connection clip lets you remove the lower part in just 2 steps. The secondary adjustment gives you greater accuracy for specific liquid types.
Safe, robust and ultralight pipettor is made from Fortron®, which is resistant to acids, alkalies, mildew, bleaches,aging, sunlight,heat, and abrasion. Fully autoclavable for simple decontamination.