Environmental Express Vacuum Manifold, 12-Pl
Representative image only.
Environmental Express Item # EW-53205-19
Use to process up to 12 FlipMate filtrations at the same time
  • PVC construction makes it resistant to most acids and other common laboratory chemicals
$500.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
Key Features
  • PVC construction makes it resistant to most acids and other common laboratory chemicals
More About this Item
The FlipMate Vacuum Manifold provides a convenient way to filter multiple samples at one time. This 12-place manifold is compatible with both the 50mL and 100mL FlipMate devices. Easily accessible valves allow for precise vacuum control during filtration, or closing of ports not in use.
One end of the manifold is equipped with a 1/4" barbed fitting to connect tubing to your vacuum pump. The other end is plugged, but the plug may be removed to connect multiple manifolds to one pump.