Environmental Express Starter Kit, Ultimate Mold Sampling; 120V
Representative image only.
Environmental Express Item # EW-60062-09
Quickly and easily sample for mold
  • Convenient kits include everything you need to check for presence of mold
  • Ultimate kit for air, surface, and inner wall sampling for mold
  • 120 VAC operation
  • 1 year warranty
PRODUCT OPTIONS
$670.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Flow Rate (LPM)30
  • Included Accessories50 Air-O-Cell® cassettes, 50 Bio-Tape™ slides, 50 mold swabs, and 10 inner wall attachments
  • Power (VAC)120
  • DescriptionStarter Kit, Ultimate Mold Sampling; 120 VAC
  • Warranty1 year
1 Year Warranty
Includes
IAQ air sampling pump with flowmeter/regulator valve and power cord with US standard plug, tubing, stand, bioisolation filter, carrying case, 50 Air-O-Cell® cassettes, 50 Bio-Tape™ slides, 50 mold swabs, and 10 inner wall attachments.
Key Features
  • Convenient kits include everything you need to check for presence of mold
  • Ultimate kit for air, surface, and inner wall sampling for mold
  • 120 VAC operation
  • 1 year warranty
More About this Item
Each starter kit includes a Z-Lite™ IAQ sampling pump with flowmeter/regulator valve and power cord with plug (US standard for 120 VAC; European for 230 VAC), tubing, stand, bioisolation filter, carrying case, plus sampling media. Select from basic, deluxe, or ultimate kit depending on your sampling needs.
Simply use the included tubing to connect the Air-O-Cell® cassettes to the Z-Lite IAQ sampling pump to collect air samples. Bio-Tape™ slides and swabs (deluxe and ultimate kits only) make it convenient to take surface samples. Connect inner wall sampling adapter (ultimate kit only) to Air-O-Cell cassette to collect samples from inside the wall. Once the sampling is complete, send the samples to a laboratory for analysis.
Included media have a limited shelf life.