Environmental Express Snip & Pour Reagents, 0.2N H2SO4, 50 x 10 mL/Pk

Representative image only.
Environmental Express Item # EW-35209-79
Deliver precise volume with pre-measured reagents
  • Easy-to-use, just snip and pour into receiver tube
  • Increase accuracy by eliminating the need to measure and pipette
  • 3 month warranty

$115.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Normality0.2
  • Empirical FormulaH2SO4
  • ClassTrapping Solution
  • FormLiquid
  • SynonymsSulfuric acid; sulphuric acid
  • Volume (mL)500
  • Weight (mL)500
  • Packaging500 mL Fifty, 10 mL PTFE ampules, per package
  • CAS Number7664-93-9
  • Description0.2N H2SO4, 50 x 10 mL/Pk
  • Warranty3 month
3 Month Warranty

Key Features

  • Easy-to-use, just snip and pour into receiver tube
  • Increase accuracy by eliminating the need to measure and pipette
  • 3 month warranty

More About this Item

Pre-measured Snip & Pour reagent tubes for Ammonia and Cyanide Distillations deliver the precise volume of reagent to each sample — working perfectly with the MicroBlock™ systems!

Snip & Pour Reagents are packaged in convenient, chemical-resistant PTFE tubes. Reagent addition is a snap with Snip & Pour reagent tubes. Just snip the corner of the PTFE tube and pour the contents directly into the receiver tube.

Each pre-measured PTFE Tube for MicroBlock delivers 10mL of reagent.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat