Environmental Express SimpleDist® Collection Traps, 25 mL; 100/Pk
Representative image only.
Environmental Express Item # EW-35209-90
Individual components for your SimpleDist system
  • Replacement manifolds and washers
  • Tubing and adapters can be replaced after wear and tear
  • Collection traps for safe pouring of the final solution
  • 3 month warranty
PRODUCT OPTIONS
$271.25USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)25
  • MaterialPolystyrene
  • Compatible WithSimpleDist Systems
  • DescriptionSimpleDist Collection Traps, 25 mL; 100/Pk
  • Warranty3 month
3 Month Warranty
Key Features
  • Replacement manifolds and washers
  • Tubing and adapters can be replaced after wear and tear
  • Collection traps for safe pouring of the final solution
  • 3 month warranty
More About this Item
Ensure cyanide and ammonia collection run smoothly by replacing parts when appropriate.
For use with SimpleDist Systems, these disposable polystyrene collection traps are clearly marked at 25mL and have a built-in vent tube that makes pouring the final solution safe and easy.