Environmental Express Hotblock Heater Mat

Representative image only.
Environmental Express Item # EW-53205-18
Keep temperatures even with the optional heating mat
  • EPA-approved hotblock systems simplify and improve metals prep and digestions
  • 1 year warranty

$657.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Length (\")24
  • Width (InchShort)12
  • Compatible WithSC174 HotPlate
  • Length (in)24
  • Width (in)12
  • Width (cm)30.48
  • Length (cm)60.96
  • Width (mm)304.8
  • Length (mm)609.6
  • DescriptionHeater Mat for Hotplate, 12x24\"
  • Warranty1 year
1 Year Warranty

Key Features

  • EPA-approved hotblock systems simplify and improve metals prep and digestions
  • 1 year warranty

More About this Item

The HotBlock® Systems provide faster, cleaner, and more cost-effective metals digestions, and are accepted for EPA Methods. Block heaters have an integrated controller and mat/block design.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat