Environmental Express GhostWipe® Moist Wipe, 15 x 15 cm, Individually Sealed; 500/Pk
Representative image only.
Environmental Express Item # EW-35208-73
Simplify your surface sample collection and preparation for lead and beryllium content
  • Just wipe surface you want to test, digest, and analyze results
  • Wipes breakdown completely and wont clog equipment
  • Complies with ASTM method E 1792 for lead or beryllium in dust
  • Each wipe is preserved in a sealed packet
  • Wipes have been used successfully on alternative elements such as Ba, Be, Cr, Cu, Se, Zn and As
  • 3 month warranty
PRODUCT OPTIONS
$142.25USD / Pkg of 500
SPECS & DESCRIPTION
Specifications & Description
  • TestSurface sampling
  • Length (\")5 29/32
  • Width (InchShort)5 29/32
  • Tests Per Kit500
  • DescriptionMoist Wipe, 15 x 15 cm, Individually Sealed; 500/Pk
  • Warranty3 month
3 Month Warranty
Key Features
  • Just wipe surface you want to test, digest, and analyze results
  • Wipes breakdown completely and wont clog equipment
  • Complies with ASTM method E 1792 for lead or beryllium in dust
  • Each wipe is preserved in a sealed packet
  • Wipes have been used successfully on alternative elements such as Ba, Be, Cr, Cu, Se, Zn and As
  • 3 month warranty
More About this Item
The Ghostwipe completely dissolves during the digestion procedure thus allowing
everything into solution. Dependent upon the material wiped, filtration may not
be necessary. In addition, the Ghostwipe is more rugged than other traditional
paper wipes and will not fall apart during the wiping of rough surfaces.
Sturdy wipe moistened with DI water can hold together even on the roughest surfaces. In the lab, they readily dissolve during the digestion process; providing better dispersions and uniform recoveries.