Environmental Express Eco-Conscious Single-Use BOD Bottles, 300 mL; 100/PK

Representative image only.
Environmental Express Item # EW-53200-02
Get more accurate results and never wash a BOD bottle again
  • Laboratory tested and EPA method 5210 B approved BOD method
  • Independent study confirms improved blanks, improved results, and improved precision
  • Reduces costs for labor and for water consumption
  • Easily identify samples with individually numbered bottles
  • Certified volume with a lower tolerance than glass (± 0.25%) to ensure accurate results
  • 3 month warranty

$134.00USD / Pkg of 100

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)300
  • MaterialPolyethylene Terephthalate (PET)
  • Bottle ColorAmber
  • Special FeaturesEPA method 5210B approved
  • Overall Height (in)5 1/2
  • Overall Width (in)2 1/2
  • DescriptionEco-Conscious Single-Use BOD Bottles, 300 mL; 100/PK
  • Warranty3 month
3 Month WarrantyEPA United States Environmental protection agency

Key Features

  • Laboratory tested and EPA method 5210 B approved BOD method
  • Independent study confirms improved blanks, improved results, and improved precision
  • Reduces costs for labor and for water consumption
  • Easily identify samples with individually numbered bottles
  • Certified volume with a lower tolerance than glass (± 0.25%) to ensure accurate results
  • 3 month warranty

More About this Item

Eco-Conscious Single-Use BOD bottles (300-mL) are lightweight, unbreakable and recyclable. They offer reduced labor, water conservation, as well as convenience. Best of all, using a fresh, clean bottle for each analysis provides more accurate results in your lab.

The PET bottles offer a special formulated carbon coating that prevents oxygen from diffusing into or out of your sample. Bottles are individually numbered. Use these bottles straight from the box without pre-cleaning. Bottles are packed in plastic- lined boxes of 100 to eliminate dust or residue that could adversely affect BOD results.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat