Environmental Express Contract Laboratory Program (CLP) Standard for ICP (4 Components) in 5% HNO3; 500 mL

Representative image only.
Environmental Express Item # EW-04573-77
Multi-component CLP standards for ICP analysis
  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3100 series
  • For AOAC Methods 990.08 and 975.03
  • CLP standard in 5% HNO3 matrix
  • 500 mL bottle

PRODUCT OPTIONS

$332.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ConcentrationMultiple
  • Matrix5% HNO3
  • Element/IonAluminum, Calcium, Magnesium, and Iron
  • FormLiquid
  • Volume (mL)500
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s. (Nitric Acid); Class 8, Packing Group II
  • Hazard SymbolCorrosive
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionContract Laboratory Program (CLP) Standard for ICP (4 Components) in 5% HNO3; 500 mL

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3100 series
  • For AOAC Methods 990.08 and 975.03
  • CLP standard in 5% HNO3 matrix
  • 500 mL bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.  

This multi-component standard contains 4 components.

• CLP standard is a 500 mL bottle which contains 5,000 µg/mL (5,000 ppm) of Aluminum, Calcium, Magnesium; and 2,000 µg/mL (2,000 ppm) of Iron in 5% HNO3.

Trace metal impurity scan is included on the Certificate of Analysis.

Hazardous material shipping fees may apply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat