EMD Millipore UFC501024 Amicon Ultra Centrifugal Filters, 0.5 mL, 10K MWCO; pack of 24

Representative image only.
EMD Millipore Mfr # UFC501024 – Item # EW-29968-22
Low-binding regenerated cellulose membrane, fast processing, and >90% recoveries
  • Unique vertical housing
  • Designed with 0.5-mL volume and 10,000 Nominal molecular weight limit (NMWL)

PRODUCT OPTIONS

$188.50USD / Pkg of 24

SPECS & DESCRIPTION

Specifications & Description

  • Min Retention Volume (µL)15
  • Tube Size (mL)0.5
  • Max Retention Volume (µL)20
  • Max Sample Volume (mL)0.5
  • Nominal Molecular Weight Limit (Mwl) (Dalton)10000
  • DescriptionAmicon Ultra Centrifugal Filters, 0.5 mL, 10K MWCO; pack of 24

Key Features

  • Unique vertical housing
  • Designed with 0.5-mL volume and 10,000 Nominal molecular weight limit (NMWL)

More About this Item

These units are premier tools for protein concentration and desalting. Typical applications include purifying macromolecular components from tissue culture extracts or cell lysates and concentrating biological samples.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat