EMD Millipore SCGPCAPRE SteriCap PLUS Bottle-Top Filter, 0.22 um PES, Sterile; 10/Pk
Representative image only.
EMD Millipore Mfr # SCGPCAPRE – Item # EW-29969-10
For ultrafast sterilization up to 20 L using vacuum filtration
  • Non-cytotoxic
  • Non-pyrogenic
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Pore Size (µm)0.22
  • Housing MaterialPolypropylene (PP)
  • SterileYes
  • DescriptionSteriCap PLUS Bottle-Top Filter, 0.22 um PES, Sterile; 10/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Non-cytotoxic
  • Non-pyrogenic
  • 1 year warranty
More About this Item
Millipore Express polyethersulfone membrane is ideal for fast sterilization of tissue culture media, serum, buffers, or other biological solutions. Model 29969-10 features a clear styrene dome and polypropylene housing. It can be used to fill any vacuum-rated bottle or other container with an inner neck diameter of 20 to 67 mm. It also has a vent port needle valve with female luer-lok™ which can be connect a 50 cc syringe filter (non included) if necessary to re-prime in the event of air locking. Model 29969-12, -14 and -16 feature styrene-acrylonitrile (SAN)/ polyethylene housing, and unique ¡°start & stop¡± function. Push a button to stop filtering, move the device to an empty bottle and restart. All units are sterile and designed for single use.