Dynalon Venting Multi-Language Labeled Safety Wash Bottle, LDPE, Ethanol, 500 mL, 5/pk

Representative image only.
Dynalon Mfr # 506495-0002 – Item # EW-62300-93
Safety phrases show in English, German, French and Spanish

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)500
  • Capacity (oz)16
  • MaterialLow Density Polyethyene (LDPE)
  • Cap ColorGreen
  • Bottle ColorTranslucent
  • Label TypeEthanol
  • AutoclavableNo
  • Outside Diameter (OD) (in)3
  • Outside Diameter (OD) (cm)7.6
  • Overall Height (in)6 1/2
  • Overall Height (cm)16.5
  • DescriptionVenting Multi-Language Labeled Safety Wash Bottle, LDPE, Ethanol, 500 mL, 5/pk

More About this Item

These wide-mouth wash bottles feature chemical and safety information indelibly printed in four languages (English, German, French, and Spanish) plus (GHS) globally harmonized symbols, NFPA diamond, and CAS number. Wash bottles feature DripLok® vapor venting valve that prevents drips. Precision jet-tip opening provides exact, accurate control. Color-coded one-piece PP closures are fitted to bottle to retain shape. Assortment packs contain one of each bottle listed.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat