DWK Life Sciences (Wheaton) W225741 Crimping Jaw Set, 8 mm, for High Performance Crimper
Representative image only.
DWK Life Sciences (Wheaton) Mfr # W225741 – Item # EW-98800-14
Interchangeable jaw sets save you money
  • Compatible with Wheaton High Performance Crimping Tool (sold separately)
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Cap Diameter (mm)8
  • DescriptionCrimping jaw set
  • Warranty1 year
1 Year Warranty
Key Features
  • Compatible with Wheaton High Performance Crimping Tool (sold separately)
  • 1 year warranty
More About this Item
Wheaton High Performance Crimping Tool (sold separately) is designed with inter-changeable jaws, so that you only need one tool and purchase the size crimper and decappers needed for your laboratory. These jaw sets are available as crimper and decappers which fit standard lab vials seals with aluminum or two-part aluminum/steel caps.