Drummond Pipet-Aid® Self-Locking Tissue Culture Filter, 0.8 µm; 5/Pk
Representative image only.
Drummond Mfr # 4-000-051 – Item # EW-14400-16
Optimize performance of your pipette controller
  • Compatible with the Drummond Pipet-Aid® Pipette Controller
  • Self-Locking, double-layer filter
PRODUCT OPTIONS
$41.50USD / Pkg of 5
Often Bought With…
SPECS & DESCRIPTION
Specifications & Description
  • ColorTranslucent
  • Compatible WithPipet-Aid® controllers
  • DescriptionPipet-Aid® Self-Locking Tissue Culture Filter, 0.8 µm; 5/Pk
Key Features
  • Compatible with the Drummond Pipet-Aid® Pipette Controller
  • Self-Locking, double-layer filter
More About this Item
Choose from a wide range of accessories and replacement parts to extend the life and optimize use of the Drummond Pipet-Aid® Pipette Controllers.
Designed and tested for use in the Pipet-Aid® controllers, tissue culture (TC) filters offer the best protection in the event over-pipetting occurs. The thin membrane in the TC filter absorbs liquid and cuts off the vacuum created by the controller, preventing the sample from contaminating, and potentially damaging, your Pipet-Aid.