Digital Barometer Data Acquisition System
Representative image only.
Item # EW-68000-51
Ideal for monitoring environmental conditions in plants and labs
  • Set high and low alarm set points
  • Connect and download real-time readings to a PC
  • Get reliable readings
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
$130.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With68000-49 (6530DS) Digital Barometer
  • DescriptionDigital Barometer Data Acquisition System
1 Year Warranty
Key Features
  • Set high and low alarm set points
  • Connect and download real-time readings to a PC
  • Get reliable readings
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
More About this Item
Use this data acquisition software and USB cable to permanently capture real-time measurements on your PC from up to eight different Traceable barometers. Data can then be saved, viewed, printed as-is, or merged into a report or spreadsheet and e-mailed to any smartphone or computer. The software also features its own 24/7 audible and visual alarm to remind you to check on data.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.