Digi-Sense Thermistor Thermometer, NIST-Traceable Calibration
Representative image only.
Digi-Sense Item # EW-20250-93
Perfect for the lab to production floor
  • Reliable measurements, guaranteed!
  • NIST-traceable certificate included
  • Heavy duty construction to protect against knocks and jars.
  • Super lager LCD for easy viewing of data
  • Accepts a wide variety of probes
  • Min, Max, and Avg readings, for quick analysis of the object
  • Electronic Offset function allows compensation of probe errors
  • 2 year warranty
$120.80USD / Each
$151.00USD / Each
Time to recalibrate?
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermistor
  • Number Of Probe Channels1
  • Types Of Probes Accepted400 Series
  • Min Temperature (° F)-40
  • Max Temperature (° F)257
  • Min Temperature (° C)-40
  • Max Temperature (° C)125
  • Accuracy±0.9⁰F (±0.5⁰C) from 32 to 158⁰F, (0 to 70⁰C); ±2⁰F (1⁰C) outside range
  • Resolution0.1⁰F (0.1⁰C)
  • Data LoggingNo
  • Low Battery IndicatorYes
  • Display TypeLarge, dual, backlit LCD
  • Battery Life3000 hours
  • Battery1 9V (included)
  • DescriptionThermistor Thermometer, NIST Traceable Calibration
  • Warranty2 year
Two year warrantyConformite EuropeenneNIST Traceable Calibration
Key Features
  • Reliable measurements, guaranteed!
  • NIST-traceable certificate included
  • Heavy duty construction to protect against knocks and jars.
  • Super lager LCD for easy viewing of data
  • Accepts a wide variety of probes
  • Min, Max, and Avg readings, for quick analysis of the object
  • Electronic Offset function allows compensation of probe errors
  • 2 year warranty
More About this Item
The Digi-Sense Professional Single-Input Thermometers offer fast response and laboratory accuracy. Features include automatic backlight, Max/Min readings, and automatic power-off. These thermometers provide precise measurements when accuracy is a must, delivering dependable results every time! Each model is packed with distinctive features for use in a wide array of applications. Robust and reliable constructed to protects against knocks and jars. These thermometers produce a high degree of accuracy, with a rapid response time, and exceptional battery life of up to 3000 hours.
Digi-Sense instrument are fully tested and calibrated to NIST-traceable standards, saving you hundreds of dollars and extra waiting time to calibrate separately.
Probe not included.