Data LoggingYes, unlimited based on smart device capacity
Memory CapacityUP to 32K IR readings
USB InterfaceYes
OutputUSB, Bluetooth®
Display TypeBacklit LCD
AlarmYes
Low Battery IndicatorYes
Battery1 9V (included)
Height (in)6
Width (in)2 1/4
Length (in)7 1/2
DescriptionProfessional Dual-Laser Infrared Thermometer with Bluetooth, 50:1
Warranty2 year
Includes
Type K flexible probe, software, tripod, carrying case, 9 V battery, and NIST-traceable calibration report with data.
Key Features
Get the most out of your data with our user-friendly mobile app
Unlimited data storage based on smart device capacity
Reliable measurements, guaranteed! Precalibrated to NIST-traceable standards
Internal data logging saves up to 32K IR readings
External type K thermocouple input for contact measurements
Dual-laser sighting to quickly pinpoint target areas
Backlight dot-matrix display permits viewing in low-light conditions
2 year warranty
More About this Item
Using our free Digi-Sense D/S IR Therm app, this dual-laser infrared (IR) thermometer with Bluetooth® technology allows you to transmit data directly to your Android or iOS device. Now you can view data at a safe distance from dangerous equipment or hazardous locations and even use your smart device as a real-time secondary display for checking measurements, eliminating the need to write down readings on paper. The results are automatically stored in the app with a date-and-time stamp and can be saved as a CSV file and emailed for future reference, manipulation, or analysis to help determine trends and conditions over a long period of time. Use the app to attach photos and notes to the records creating a clear reference point for your work—ideal for record keeping at large sites.
The IR thermometer offers fast response time and high accuracy due to its 50 to 1 distance-to-sight ratio. Dual targeting lasers provide precise indication of the measurement area. It is ideal for a wide range of applications including food preparation, safety and fire inspection, plastic molding, marine, HVAC, and fleet maintenance.
Advanced features include internal data logging, high/low temperature alarms, adjustable emissivity, temperature differential, data Hold, Max/Min/Avg readings, USB interface for data transmission, and auto power-off. The meter also supports contact measurement through a type K thermocouple input.
The instrument is fully tested and calibrated to NIST-traceable standards for reliable measurements right out of the box, saving you time and money!
Cole-Parmer provides a wide offering of Professional Infrared Thermometers with varying features and accuracies depending on your application. We now offer an FDA-cleared infrared thermometer. Our other infrared thermometers are not approved for medical usage and are not FDA-approved. For help in selecting an infrared thermometer please contact the Technical Support team at Cole-Parmer.
Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.