Digi-Sense Probe Type-J Inconl 24" L Almhd
Representative image only.
Digi-Sense Item # EW-90446-75
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeJ
  • Sheath Length (in)24
  • DescriptionProbe Type-J Inconl 24\" L Almhd
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These MgO thermocouple assemblies consist of a single or dual thermocouple element constructed of various sheath materials, assembled together with a connection head.
FEATURES
  • Inconel, 316 Stainless steel or 304 Stainless steel sheath materials
  • 1/2'' X 1/2'' NPT stainless steel hex nipple for head mounting fittings
  • Aluminum screw cover, Cast iron screw cover, or Cast iron/aluminum explosion proof class C Heads
  • Standard Limits of Error
  • Grounded Measuring Junction
  • Custom built assemblies available upon request