Digi-Sense Mini Particle Counter with Bluetooth® Connectivity

Representative image only.
Digi-Sense Item # EW-20250-65
View, store, and share real-time data on your smartphone
  • Get the most out of your data with our user-friendly mobile app
  • Unlimited data storage based on smart device capacity
  • Monitors particles, air temperature, and relative humidity
  • View measurements clearly on 2" color TFT LCD
  • Tilt stand offers easy hands-free viewing
  • 2 year warranty

$465.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Particle Size (micron)2.5, 10
  • Max Temperature (° F)122
  • Min Temperature (° F)32
  • Max Temperature (° C)50
  • Min Temperature (° C)0
  • Min Humidity (RH)0
  • Max Humidity (RH)100
  • RH Resolution (%)0.1
  • Temperature Resolution (° F)0.1°F
  • Temperature Resolution (° C)0.1°C
  • Temperature Accuracy±2°F/1°C
  • Humidity Accuracy±3.5% RH from 20 to 80% RH; ±5% RH remaining
  • Display2.0’’ TFT Color LCD
  • Memory5000 data sets and unlimited based on smart device capacity
  • OutputUSB, Bluetooth®
  • Power3.5 V Li-Ion
  • Height (in)7.25
  • Depth (in)1.5
  • Width (in)2.125
  • Height (cm)18.5
  • Depth (cm)3.8
  • Width (cm)5.5
  • DescriptionMini Particle Counter with Bluetooth®
  • Warranty2 year
Two year warrantyApple App StoreConformite EuropeenneGoogle Play App

Includes

Kick stand, protective cap, and one 9 V battery.

Key Features

  • Get the most out of your data with our user-friendly mobile app
  • Unlimited data storage based on smart device capacity
  • Monitors particles, air temperature, and relative humidity
  • View measurements clearly on 2" color TFT LCD
  • Tilt stand offers easy hands-free viewing
  • 2 year warranty

More About this Item

Using our free Digi-Sense D/S Particle app, this mini particle counter with Bluetooth® technology allows you to transmit data directly to your Android or iOS device. Now you can view data at a safe distance from dangerous equipment or hazardous locations and even use your smart device as a real-time secondary display for checking measurements, eliminating the need to write down readings on paper. The results are automatically stored in the app with a date-and-time stamp and can be saved as a CSV file and emailed for future reference, manipulation, or analysis to help determine trends and conditions over a long period of time. Use the app to attach photos and notes to the records creating a clear reference point for your work—ideal for record keeping at large sites.

The handheld particle counter provides fast, easy, and accurate readings for particulate matter and particle mass concentration with air temperature and relative humidity measurements. It allows you to simultaneously measure PM2.5 and PM10. Other features include a selectable sample time, analog bar indicator, °F/°C selection, clock display, and auto power-off. Ideal for environmental protection and energy-saving applications.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat