Digi-Sense K20-2-350 Thermocouple Wire Type K 20-Gauge, Ceramic Fiber Braid Insulation 08541-24
Representative image only.
Digi-Sense Item # EW-08541-24
Make your own extension cables
  • 6 month warranty
PRODUCT OPTIONS
$560.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialCeramic Fiber
  • Wire Gauge (Ga)20
  • Error Limit±2% reading from -330 to -165°F (-200 to -110°C); ±4°F (±2.2°C) from -165 to 545°F (-110 to 285°C); ±0.75% reading from 545 to 2300°F (285 to 1250°C)
  • Inner InsulationCeramic Fiber Braid
  • Outer InsulationCeramic Fiber Braid
  • Stranded / SolidSolid
  • OverbraidNo
  • Min Temperature (° F)0
  • Max Temperature (° F)2200
  • Min Temperature (° C)-18
  • Max Temperature (° C)1427
  • Length (ft)100
  • Size (in)0.092 in x 0.154 in
  • DescriptionThermocouple Wire Type K 20-Gauge, Ceramic Fiber Braid Insulation, 100-ft Spool
  • Warranty6 month
6 Month Warranty
Key Features
  • 6 month warranty
More About this Item
Wire comes in 20-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.