Diba Omnifit® Bottle Thread Adapter, S40 (Female) to GL45 (Male), PTFE
Representative image only.
Diba Mfr # 00945A-S40 – Item # EW-21942-99
Optimize performance of your solvent safety systems
  • Constructed of chemically inert PTFE
$78.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBottle Thread Adapter, S40 (female) to GL45 (male); 1/ea
Key Features
  • Constructed of chemically inert PTFE
More About this Item
Bottle thread adapter converts bottles with S40 threads to be use with any GL45 thread caps.