DeFelsko PosiTest AT50 Digital Adhesion Tester 50-mm Positest Accessory Kit
Representative image only.
DeFelsko Mfr # AT50KIT – Item # EW-59785-15
Interchangeable standoffs allow adhesion testers to pull 50 mm dollies
  • 1 year warranty
$300.00USD / Each
$166.50 - $239.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAdhesion Tester 50-mm Positest Conversion Kit
  • Warranty1 year
1 Year Warranty
Includes
50 mm standoff, 50 mm hole saw, and twelve 50 mm aluminum test dollies.
Key Features
  • 1 year warranty
More About this Item
Interchangeable stand-off allows a PosiTest ATM20 or ATA20 to pull 50 mm dollies. Larger surface area of dolly provides improve low rang precision and repeatability. Ideal for lower bond strength finishes such as coatings on wood, concrete, and plastic. To be used with the PosiTest ATM Manual and PosiTest ATA Automatic Adhesion Testers.