DeFelsko PosiPen Posipen A Coating Thickness Gauge/Pen, 0.25-20 mils
Representative image only.
DeFelsko Mfr # POSIPEN A – Item # EW-59785-18
Ideal for small, hot or hard-to-reach surfaces
  • Lightweight—easy to carry, just like a ball point pen
  • Rugged —not affected by mechanical shock, acid, oil, water and dust
  • No user adjustment required
  • Highly wear resistant probe tip for long life and continuous accuracy
  • 1 year warranty
$250.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Range0.25 to 20 mil (5 to 500 µm)
  • ScaleFerrous/Nonferrous
  • DescriptionPosipen A Coating Thickness Gauge/Pen, 0.25-20 mils
  • Warranty1 year
1 Year Warranty
Includes
Leather pouch, instructions, and long form Certificate of Calibration traceable to PTB.
Key Features
  • Lightweight—easy to carry, just like a ball point pen
  • Rugged —not affected by mechanical shock, acid, oil, water and dust
  • No user adjustment required
  • Highly wear resistant probe tip for long life and continuous accuracy
  • 1 year warranty
More About this Item
PosiPen has a very small, unique magnet which can be placed with pin-point accuracy on extremely small parts, and on peaks and valleys. Ideal for measuring non-magnetic coatings such as paint, enamel, plating, and hot-dip galvanizing on steel.
The pen features a triple indicator. Use the red/silver joining line when the gauge is horizontal (walls). Use the green line when the gauge is pointing straight down (green/ground). Use the blue line when the gauge is pointing straight up (blue/sky).