CPS Products TM360 Temp-Seeker Multi-Port Thermo-psychrometer
Representative image only.
CPS Products Mfr # TM360 – Item # EW-10110-89
Upgradeable to measure relative humidity, dew point and dry bulb using the CPTMX2RH accessory probe
  • 4 remote temperature stations
  • Extreme accuracy up to 0.2degF/.1degC Unique SCAN function
  • Large probe storage area
  • Min/Max measurements
  • Memory feature
  • degF/degC selectable
  • 1 year warranty
$396.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeGlass
  • Resolution0.2°F (0.1°C)
  • DescriptionTemp-Seeker Multi-Port Thermo-psychrometer
  • Warranty1 year
1 Year Warranty
Key Features
  • 4 remote temperature stations
  • Extreme accuracy up to 0.2degF/.1degC Unique SCAN function
  • Large probe storage area
  • Min/Max measurements
  • Memory feature
  • degF/degC selectable
  • 1 year warranty
More About this Item
The TEMP-SEEKER® is a professional grade temperature measurement instrument with 4 remote stations and a wide range of job specific accessory probes to complete all of your temperature testing needs. Measures temperature differential between any two of the four remote stations and incorporates a unique Scan function that alternates measurement readings between all four of its remote stations in three-second intervals.