CPS Products HS3 Standard Refrigerant Charge Hose, 3 ft/90 cm
Representative image only.
CPS Products Mfr # HS3 – Item # EW-10111-01
1/4" Standard charging hoses
  • All-weather flexibility and strength
  • Extra deep knurling on nuts for easy grip
  • Available with MicroSeal™ or ball valve fittings
  • Leak-proof eight-sided crimp
  • Blow-off grooves channel refrigerant to prevent spray back
  • 1 year warranty
$39.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionStandard Refrigerant Charge Hose, 3 ft/90 cm
  • Warranty1 year
1 Year Warranty
Key Features
  • All-weather flexibility and strength
  • Extra deep knurling on nuts for easy grip
  • Available with MicroSeal™ or ball valve fittings
  • Leak-proof eight-sided crimp
  • Blow-off grooves channel refrigerant to prevent spray back
  • 1 year warranty
More About this Item
The hose has 1/4" SAE fittings with a 45° bend on one end. It can operate at 750 psi working pressure with a burst pressure of 3000 psi.