Cole-Parmer Essentials Lid for 96-Place Reversible Tube Rack, PP, Translucent; 5/PK
Representative image only.
Cole-Parmer Essentials Item # EW-06736-11
Withstand temperatures down to -80 °C (-112 °F)
  • Autoclavable
  • 1 year warranty
PRODUCT OPTIONS
$20.70USD / Pkg of 5
BUY MORE & SAVE
  • 1-3$20.70USD / Pkg of 5
  • 4-19$18.63USD / Pkg of 5
  • 20-79$17.60USD / Pkg of 5
  • 80+$16.56USD / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)8 3/8
  • Width (in)4 1/2
  • Height (in)1 1/16
  • MaterialPolypropylene(PP)
  • ColorTranslucent
  • DescriptionCover for 96-well microcentrifuge tube racks
  • Warranty1 year
3 Month Warranty
Key Features
  • Autoclavable
  • 1 year warranty
More About this Item
One side of this polypropylene (PP) racks is designed to hold 0.5mL microcentrifuge tubes; the other side holds 1.5 to 2.0mL tubes. Withstand temperatures down to -80°C (-112°F).