Coupling hardware for 5/8" diameter bore, 7.2 hp maximum
Representative image only.
Item # EW-07127-59
Create your own motor mounting with the hardware below
  • Optimize performance of your Pump Motor
$22.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Motor hp7.2
  • DescriptionCoupling hardware for 5/8" diameter bore, 7.2 hp maximum
Key Features
  • Optimize performance of your Pump Motor
More About this Item
Order one coupling with the same diameter as the motor driving shaft, one coupling with the same diameter as the driven shaft, and one spider (see accessories).