Costar Spin-X 8163 Centrifuge Tube Filters, Spinfilter Ca 0.45µm 100/cs
Representative image only.
Costar Spin-X Mfr # 8163 – Item # EW-01937-36
Membrane-containing filter unit within a centrifuge tube
  • Removing bacteria, cells, and particles from liquids
  • DNA removal from agarose or acrylamide gels
PRODUCT OPTIONS
$252.50USD / Case of 100
SPECS & DESCRIPTION
Specifications & Description
  • MembraneCellulose Acetate
  • Pore Size (µm)0.45
  • Working Volume (µL)500
  • Tube Size (mL)2
  • DescriptionCentrifuge Tube Filters, Spinfilter Ca 0.45µm 100/cs
Key Features
  • Removing bacteria, cells, and particles from liquids
  • DNA removal from agarose or acrylamide gels
More About this Item
Ideal for HPLC sample preparation.