Costar 96-Well Microplate, Deep Well, Sterile, Round Bottom, 1 mL; 25/Cs
Representative image only.
Costar Mfr # 3958 – Item # EW-01729-02
Uniform skirt heights for greater robotic gripping surface
  • Constructed of clarified polypropylene for optimal clarity and solvent resistance
  • Certified RNase and DNase free
PRODUCT OPTIONS
$250.00USD / Case of 25
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (mL)1
  • Well ShapeRound
  • Surface TreatmentUntreated
  • Lid (Included)No
  • SterileYes
  • Plate TypeDeep Well
  • Plate ColorClear
  • MaterialPolypropylene
  • Packaging5 Plates/Pack, 25 Plates/Case
  • Description96-Well Microplate, Deep Well, Sterile, Round Bottom, 1 mL; 25/Cs
Key Features
  • Constructed of clarified polypropylene for optimal clarity and solvent resistance
  • Certified RNase and DNase free
More About this Item
Standard microplates are built to industry standard footprint and compatible with automated systems. Deep-well plates are ideal for higher-volume storage and assay applications. Lids are not included, for standard microplates and 1.0-mL deep well plates, use storage mat 01729-22 and applicator 01729-24 sold separately.