Costar 96-Well Microplate, Standard Well, Non-Sterile, Flat Well, 0.36 mL; 100/Cs

Representative image only.
Costar Mfr # 3364 – Item # EW-01728-81
Uniform skirt heights for greater robotic gripping surface
  • Constructed of clarified polypropylene for optimal clarity and solvent resistance
  • Certified RNase and DNase free

PRODUCT OPTIONS

$425.00USD / Case of 100

SPECS & DESCRIPTION

Specifications & Description

  • Number of Wells96
  • Well Volume (mL)0.36
  • Min Working Well Volume (mL)0.075
  • Max Working Well Volume (mL)0.2
  • Well ShapeFlat
  • Surface TreatmentUntreated
  • Lid (Included)No
  • SterileNo
  • Plate TypeStandard Well
  • Plate ColorClear
  • MaterialPolypropylene
  • Packaging25 Plates/Pack, 100 Plates/Case
  • Description96-Well Microplate, Standard Well, Non-Sterile, Flat Well, 0.36 mL; 100/Cs

Key Features

  • Constructed of clarified polypropylene for optimal clarity and solvent resistance
  • Certified RNase and DNase free

More About this Item

Standard microplates are built to industry standard footprint and compatible with automated systems. Deep-well plates are ideal for higher-volume storage and assay applications. Lids are not included, for standard microplates and 1.0-mL deep well plates, use storage mat 01729-22 and applicator 01729-24 sold separately.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat