Costar 3363 96-Well Standard Microplates, Nontreated, PP, V-Bottom
Representative image only.
Costar Mfr # 3363 – Item # EW-01728-80
96-Well Microplates and Assay Blocks, Nontreated, PP
  • Constructed of clarified polypropylene for optimal clarity and solvent resistance—ideal for compound library and combinatorial chemistry applications
PRODUCT OPTIONS
$364.00USD / Case of 100
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (mL)0.32
  • Well ShapeV-bottom
  • Surface TreatmentUntreated
  • Plate TypeStandard microplate
  • MaterialPolypropylene
  • Description96-Well Standard Microplates, Nontreated, PP, V-Bottom
Key Features
  • Constructed of clarified polypropylene for optimal clarity and solvent resistance—ideal for compound library and combinatorial chemistry applications
More About this Item
Standard microplates are built to industry standard footprint; compatible with automated systems. Plates are certified RNase- and Dnase-free.