Corning 432076 CoolCell® Filler Vials, 2 mL; 6/Pk
Representative image only.
Corning Mfr # 432076 – Item # EW-04392-76
No need to freeze a full load of samples
  • Ensure cell freezing rate consistency and uniform results
  • 1 year warranty
PRODUCT OPTIONS
$35.00USD / Pkg of 6
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)2
  • Compatible WithCoolCell LX, CoolCell FTS30 and CoolCell 5ml LX containers
  • DescriptionFiller Vials, 2 mL; 6/pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Ensure cell freezing rate consistency and uniform results
  • 1 year warranty
More About this Item
Use filler vials when you need to freeze less than a full load of samples to maintain a consistent cooling profile. To ensure cell freezing rate consistency and uniform results when using CoolCell containers, insert a CoolCell Filler Vial into empty wells when freezing less than a full load.