Corning 3495 Permeable Supports, 24-ClusterPlate/0.4um/PTFE; 24/Cs
Representative image only.
Corning Mfr # 3495 – Item # EW-19480-23
Essential tool for the study of both anchorage-dependent and independent cell lines
  • Provide an environment for cells that closely resembles their in vivo state
  • Treated for optimal cell attachment
  • Ideal for a variety of cell culture applications—chemotaxis, drug transport, tissue remodeling, and invasion studies
  • Supports come sterile and preloaded in cluster plates
  • Sterilized by gamma radiation
SPECS & DESCRIPTION
Specifications & Description
  • Cluster Plate Size Number24
  • Well Diameter (mm)6.5
  • Well ShapeFlat
  • Culture Area (cm²)0.3
  • Surface TreatmentTC-treated
  • Lid (Included)Yes
  • SterileYes
  • Membrane MaterialPTFE
  • DescriptionPermeableSupports,24-ClusterPlate/0.4um/PTFE;24/Cs
Key Features
  • Provide an environment for cells that closely resembles their in vivo state
  • Treated for optimal cell attachment
  • Ideal for a variety of cell culture applications—chemotaxis, drug transport, tissue remodeling, and invasion studies
  • Supports come sterile and preloaded in cluster plates
  • Sterilized by gamma radiation
More About this Item
These permeable supports, also known as cell culture inserts, are designed to satisfy your diverse cell culture requirements. A suspended design prevents media wicking between the insert and the plate well. It also allows for co-culturing without damaging cells attached to the plate's growth surface. Choose either polycarbonate (PC) or Polyethylene Terephthalate (PET) membrane material. Polycarbonate membranes are translucent while PET membranes are clear, allowing for excellent visibility under phase contrast microscopy.