Corning 3081 96-Well Plate/Block Storage Mat Applicator
Representative image only.
Corning Mfr # 3081 – Item # EW-01729-24
Tightly seals microplate wells to prevents spills and well-to-well contamination
  • Manufactured from EVA (ethyl vinyl acetate) and polymer is compatible with DMSO
  • Can be applied manually or with our Storage Mat Applicator
SPECS & DESCRIPTION
Specifications & Description
  • Description96-Well Plate/Block Storage Mat Applicator
Key Features
  • Manufactured from EVA (ethyl vinyl acetate) and polymer is compatible with DMSO
  • Can be applied manually or with our Storage Mat Applicator
More About this Item
Storage mat seals each well individually for long-term storage—virtually eliminates moisture loss. Use storage mats with standard microplates and 1.0-mL deep-well assay blocks.
Applicator features a sturdy, comfortable handle and an adapter for plates of different heights.