Corning Stripwell™ Strip Holder, 96-Well; 20/Cs
Representative image only.
Corning Mfr # 2572 – Item # EW-13020-50
Designed for use with Corning® Stripwell™ strips and plates
  • Constructed of durable polystyrene
  • Holds twelve 1 x 8 strips
  • Allows each individual well to be positioned back into the plate once broken apart
PRODUCT OPTIONS
$170.00USD / Case of 20
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Lid (Included)No
  • Plate TypeHolder
  • MaterialPolystyrene
  • Height (in)9/16
  • Width (in)3 3/8
  • Length (in)5 1/40
  • DescriptionStrip Holder, 96-Well; 20/Cs
Key Features
  • Constructed of durable polystyrene
  • Holds twelve 1 x 8 strips
  • Allows each individual well to be positioned back into the plate once broken apart
More About this Item
These accessories are compatible with the Corning® Stripwell™ flat bottom strips and 96-well plates. The strip holder is the 96-well egg crate style holder for 1 x 8 Stripwell™ strips, model 13020-12. The strip ejector is designed to remove Stripwell™ strips from the egg crate holder. Strips and plates sold separately.