Cole-Parmer WP1BB97P Fibrous polypropylene string-wound cartridge, 1 µm, 10" length
Representative image only.
Cole-Parmer Item # EW-01508-77
String-wound pattern provides greater surface area for lower pressure drop and higher flow
PRODUCT OPTIONS
$33.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene
  • Micron Rating1
  • Filter Length (in)10
  • DescriptionFibrous polypropylene string-wound cartridge, 1 µm, 10" length
More About this Item
Polypropylene string is resistant to chemical and bacterial attack. Suitable to 165°F (74 °C).
Fibrous polypropylene cartridges have a lower pressure drop than fibrillated cartridges but use an antistatic agent that may cause foaming in some applications.