Cole-Parmer WP.5BB97P Fibrous polypropylene string-wound cartridge, 0.5 µm, 10" length
Representative image only.
Cole-Parmer Item # EW-01508-78
String-wound pattern provides greater surface area for lower pressure drop and higher flow
PRODUCT OPTIONS
$38.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene
  • Micron Rating0.5
  • Filter Length (in)10
  • DescriptionFibrous polypropylene string-wound cartridge, 0.5 µm, 10" length
More About this Item
Polypropylene string is resistant to chemical and bacterial attack. Suitable to 165°F (74 °C).
Fibrous polypropylene cartridges have a lower pressure drop than fibrillated cartridges but use an antistatic agent that may cause foaming in some applications.