Cole-Parmer Water Vapor Permeation Determination Kit; 1/Ea
Representative image only.
Cole-Parmer Item # EW-10100-99
  • For use with Cole-Parmer moisture determination balances
  • Shorten permeability test time from 72 hours to just 50 minutes
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeWater Vapour Determination Kit
  • DescriptionWater Vapor Permeation Determination Kit; 1/Ea
1 Month Warranty
Key Features
  • For use with Cole-Parmer moisture determination balances
  • Shorten permeability test time from 72 hours to just 50 minutes
  • 1 month warranty
More About this Item
Replacement parts and accessories for your Cole-Parmer Basic and Advanced Touch-Screen Balances.
The set is used for determination of water vapour permeability within various materials such as cloth, knitted fabric, or leather. Permeability of water vapour is a feature which directly determines quality and application possibility of specific materials. Utilizing this set with the Symmety moisture determination balance shortens the procedure time from 72 hours to just 50 minutes!