Cole-Parmer Two-Stage Regulator, 1200 scfh Capacity, 350 CGA Fitting
Representative image only.
Cole-Parmer Item # EW-98202-32
  • 1 year warranty
PRODUCT OPTIONS
$608.60USD / Each
$716.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Delivery Pressure (psig)100
  • Max Inlet Pressure (PSI)3000
  • Outlet Connections¼\" NPT(M)
  • CGA Fitting540
  • Capacity (SCFH)1200
  • Outlet Gauge Graduations (PSI)5
  • Inlet Gauge Graduations (PSI)100
  • DescriptionTwo-Stage Regulator, 1200 scfh Capacity, 350 CGA Fitting
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Two-stage regulators reduce cylinder pressure in two stages. Use first stage to approach working pressure; use second stage to permit precise adjustment or working pressure. Second stage is overload protected.